Order Entry
Canada
Orders LinkContactUsLinkComponent
Ward's® Electrophoresis Techniques Reference Poster

Ward's® Electrophoresis Techniques Reference Poster

Electrophoresis Techniques Reference Poster, 38", 27"
Catalog # 470023-282
Supplier:   Avantor
Ward's® Electrophoresis Techniques Reference Poster
Catalog # 470023-282
Supplier:  Avantor
Ward's® Electrophoresis Techniques Reference Poster
Ward's® Electrophoresis Techniques Reference Poster

Electrophoresis Techniques Reference Poster, 38", 27"

Price
47,99 $
Log in for availability
Each
Product Details & Documents

Clearly Presents Guidelines for Agarose and PAGE Procedures

  • All key steps and applications of gel electrophoresis
  • Excellent wall reference
  • Artfully illustrated

Students will always know which step comes next when our exclusive poster is on display in the lab. Agarose electrophoresis for DNA analysis and polyacrylamide gel electrophoresis (PAGE) for protein analysis are both detailed in a step-by-step format, highlighted with photographs and illustrations that show how to cast, load, run, stain, and distain a gel. It also features information on the use of restriction enzymes, protein sample preparation, preparation of gels, running buffers, and the use of DNA and protein stains, as well as equipment needed.

Specifications
  • Width:
    38"
  • Description:
    Electrophoresis Techniques Reference Poster
  • Height:
    27"