ContactUsLinkComponent
Economy Rod-Mounted Pulley
Economy Rod-Mounted Pulley
Catalog # 470149-504
undefined
Economy Rod-Mounted Pulley
Catalog # 470149-504
Supplier Number:  P10317A
Specifications
  • Description:
    Economy Rod-Mounted Pulley
  • Environmentally Preferable:
  • Cat. no.:
    470149-504
  • Supplier No.:
    P10317A
Specifications
About this item
Excellent for Introductory Mechanics Studies
  • Low friction cone bearings ensure accurate, repeatable results
Consists of a 10cm-long rod securely fastened to a durable nylon fork. The nylon sheave is 4.5cm in diameter.