Order Entry
Canada
ContactUsLinkComponent
 
Microjaw Extension Clamp
  470039-378
undefined
Microjaw Extension Clamp
  470039-378
 :  ECLJ03

 

  • Description:
    Microjaw Extension Clamp
  • Length:
    10"
  • Cat. no.:
    470039-378
  • Clamp Extension:
    4"
  • Supplier No.:
    ECLJ03

 

 

Long 10" extension clamp with three prongs.

  • Rubber coated prongs
  • Stainless steel rod

One vinyl-coated jaw is fixed, while the other opens gradually against a stainless steel compression spring without binding. The corrosion-resistant, easy-to-clean, high-strength alloy withstands high temperatures. Length: 10". Maximum jaw opening: 4".