Orders LinkContactUsLinkComponent
Environmentally Friendly Thermometers
Supplier:   Thermco
Environmentally Friendly Thermometers
Supplier:  Thermco
Certificates
Environmentally Friendly Thermometers
Environmentally Friendly Thermometers
4 Options Available Below
Product Details & Documents
Glass thermometers with partial or total immersion.
Product Family Options
Product Information
  • Range
    Length
    Availability
    Price
  • 470019-642
    Environmentally Friendly Thermometers, 1 °C, Partial Immersion
    Environmentally Friendly Thermometers, 1 °C, Partial Immersion
    −20...+110 °C
    12"
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $33.20
    $33.20
    Log in for availability
    Box of 10
    Box of 10 - $249.65
    $249.65
    Each
       $33.20
    Box of 10
       $249.65
    Supplier Number: B110CY3BLK
  • 470019-644
    Environmentally Friendly Thermometers, 1 °C, Total Immersion
    Environmentally Friendly Thermometers, 1 °C, Total Immersion
    −20...+110 °C
    12"
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $33.20
    $33.20
    Log in for availability
    Box of 10
    Box of 10 - $249.65
    $249.65
    Each
       $33.20
    Box of 10
       $249.65
    Supplier Number: B110CYBLK
  • 470019-646
    Environmentally Friendly Thermometers, 1 °C, Partial Immersion
    Environmentally Friendly Thermometers, 1 °C, Partial Immersion
    −20...+150 °C
    12"
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $33.20
    $33.20
    Log in for availability
    Box of 10
    Box of 10 - $251.20
    $251.20
    Each
       $33.20
    Box of 10
       $251.20
    Supplier Number: B150CY3BLK
  • 470019-648
    Environmentally Friendly Thermometers, 1 °C, Total Immersion
    Environmentally Friendly Thermometers, 1 °C, Total Immersion
    −20...+150 °C
    12"
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $12.00
    $12.00
    Clearance
    Log in for availability
    Box of 10
    Box of 10 - $120.10
    $120.10
    Clearance
    Each
       $12.00
    Box of 10
       $120.10
    Supplier Number: B150CYBLK