Dynamo
470120-948
: Wards
- Description:Dynamo
- Environmentally Preferable:
- Cat. no.:470120-948
- Supplier No.:PH1245-N8
The Dynamo illustrates how electricity is produced.
- Hand-Wheel Apparatus Generates Current
- Two Lamp Holders and Bulbs Represent Currents
- Wooden Base Attached to Apparatus
- Process Shows How Electricity Is Produced
The hand-wheel apparatus generates alternating and direct current.
Two lamp holders and bulbs represent currents, and the apparatus is attached to a polished wooden base.