Duodenum Slide
Duodenum, Cross section, Hematoxylin and Eosin
470177-614
: Avantor
Explore a section of the digestive system
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. Shows Brunner’s glands and intestinal glands, known as Lieberkuhn’s crypts.
- Description:Duodenum
- Stain:Hematoxylin and Eosin
- Orientation:Cross section