Ward's® Drosophila Vial Set
Catalog #: 470016-942
Supplier: Avantor
Specifications
- Cap included:Yes
- Cap Type:Plug
- Description:Drosophila Vial Set
- Cat. no.:470016-942
- Supplier No.:WARD470016-942
Specifications
About this item
This set includes specially designed foam plugs.
- Plugs ensure adequate ventilation
Vials and plugs prevent escape.
Packaging: One package has ten sets.
Customers Who Bought This Also Bought