You searched for: Tissue Grinders
Tissue grinders efficiently crush samples using a method that eliminates the possibility personnel will come in contact and contaminate contents. Available in different strength levels, sizes, and abrasiveness, any material being handled can be reduced in volume or particle size. While thin grinders are ideal for soft organic homogenates, fibrous samples require more powerful tools for enhanced grinding action. The tissue grinders will properly prepare samples for a homogenized uniformity.
VWR® Disposable Tissue Grinders
Supplier: VWR International
These disposable tissue grinders are safe and easy to use.
Expand 3 Items
VWR® Tissue Grinders, PTFE
Supplier: VWR International
Composed of a stainless steel plunger with pure PTFE head and a precision borosilicate glass vessel.
Expand 8 Items
VWR® Disposable Pellet Mixers and Cordless Motor
Supplier: VWR International
Designed to homogenize cells and plant tissue or to re-suspend protein and DNA pellets.
Expand 7 Items
VWR® Ultra Tissue Grinders, Sterile
Supplier: VWR International
Vitrified silica-cast pestle tips provide enhanced grinding action for difficult and fibrous samples such as muscle or tissue with bone fragments.
Expand 2 Items
Interchangeable Tissue Grinder, Ace Glass
Supplier: Ace Glass
This is a precision ground instrument for macerating tissue
Expand 3 Items
Dura-Grind™ Stainless Steel Dounce Tissue Grinder, Wheaton®
Supplier: DWK Life Sciences (KIMBLE)
Wheaton Dura-Grind™ stainless steel Dounce tissue grinders provide a rugged alternative to glass Dounce tissue grinders.
Expand 2 Items
KIMBLE® Microscale Tissue Grinder Kit with Grinder and Pestle Varieties, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Kit contains a variety of the finest microscale tissue grinders available. It includes ground glass, smooth glass and PTFE grinders suitable for tough connective tissue and soft tissue such as brain or liver.
Expand 1 Items
Micro Tissue Grinder with Cap, Wheaton
Supplier: DWK Life Sciences (KIMBLE)
Grinders for preparation of soft tissue homogenates.
Expand 1 Items
Dounce Tissue Grinders, Wheaton
Supplier: DWK Life Sciences (KIMBLE)
Designed for fine particle size reduction without damage to cell nuclei
Expand 4 Items
Micro-Tissue Grinder Kit, Wheaton
Supplier: DWK Life Sciences (KIMBLE)
Micro tissue grinder kit is ideal for sterile homogenization processes.
Expand 1 Items
KIMBLE® Micro DUALL® Tissue Grinders, All-Glass with Screw-Cap, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Serve as a storage vessel following homogenization. Centrifugation, concentrations and lyophilization can be accomplished without a transfer.
Expand 4 Items
Micro Tissue Grinder, Disposable, Wheaton
Supplier: DWK Life Sciences
The Disposable Micro Tissue Grinder allows for homogenized tissue micro samples without cross-contamination
Expand 1 Items
Tissue Homogenizer, Dounce, Ace Glass Incorporated
Supplier: Ace Glass
This Dounce tissue homogenizer is supplied with two pestles, one "loose" and one "tight", to ensure dissociation of cells into fine particles with minimal damage to cell nuclei
Expand 4 Items
PYREX® Potter-Elvehjem Tissue Grinder with Fluoropolymer Resin Pestles, Corning
Supplier: Corning
Homogenizers designed for preparing small samples for pathological and biochemical studies.
Expand 7 Items
Micro Tissue Grinder with PTFE Spacer, Wheaton
Supplier: DWK Life Sciences (KIMBLE)
PTFE spacer aligns the pestle and reduces aerosoling of the product.
Expand 1 Items
KIMBLE® Potter-Elvehjem Tissue Grinder with PTFE Pestle and Glass Tube, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Large volume tissue grinder with a finely machined PTFE pestle.
Expand 1 Items
Safe-Grind® Plastic-Coated Tissue Grinders with Fluoropolymer Resin Pestles, Wheaton
Supplier: DWK Life Sciences (KIMBLE)
For sure grip and safe grinding of homogenate even with power homogenization.
Expand 5 Items
Tapered Tissue Grinders, Wheaton
Supplier: DWK Life Sciences (KIMBLE)
Grinders are available with glass or fluoropolymer resin pestles
Expand 7 Items
Tenbroeck Tissue Grinders, Wheaton
Supplier: DWK Life Sciences (KIMBLE)
For homogenizing delicate tissues such as intestines, liver, and heart
Expand 5 Items
Tissue Homogenizer, Tenbroeck, Ace Glass Incorporated
Supplier: Ace Glass
These all-glass Tenbroeck tissue grinders come with precision-made, interchangeable pestles and tubes
Expand 3 Items
CS1 and CS2 Sterile Tissue Grinders, Kimble Chase
Supplier: Kimble Chase
CS1 and CS2 Sterile Tissue Grinders are easy to use and designed for your safety.
Expand 3 Items
KIMBLE® DUALL® Tissue Grinders, with PTFE Pestles and Glass Tube, with Screw Cap, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
The DUALL® is an extremely efficient grinder which combines both conical and cylindrical surfaces on a single pestle and tube. These separate areas perform the dual functions of initial grinding in the conical section and final homogenization in the cylindrical section.
Expand 2 Items
Potter-Elvehjem Tissue Grinders, Wheaton
Supplier: DWK Life Sciences (KIMBLE)
Grinders for preparation of soft tissue homogenates, such as liver, brain.
Expand 19 Items
Tissue Homogenizer, Potter-Elvehjem, Micro, Ace Glass Incorporated
Supplier: Ace Glass
This micro size tissue homogenizer is for extremely precise work
Expand 1 Items
Tissue Homogenizer, Potter-Elehjem, Ace Glass Incorporated
Supplier: Ace Glass
When using this Potter-Elehjem tissue homogenizer, homogenization occurs as the sample and buffer are forced through the cylindrical portion of the mortar as the pestle is rotated downward
Expand 6 Items
Tissue Homogenizer, Ace Glass Incorporated
Supplier: Ace Glass
This highly efficient, all glass, tapered tissue grinder/homogenizer combines two grinding surfaces on a single pestle and tube
Expand 12 Items
KONTES® Potter-Elvehjem Tissue Grinders with PTFE Pestles, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
For homogenization of tissue samples
Expand 7 Items
KIMBLE® DUALL® Tissue Grinders, PTFE Pestles and Glass Tubes, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Tissue grinders greatly improve grinding efficiency by combining both conical and cylindrical surfaces to effectively reduce tissue and produce a uniform homogenate
Expand 4 Items
SP Wilmad-LabGlass Dounce Tissue Grinders, SP Industries
Supplier: SP Industries
Dounce tissue grinders are made from smooth surfaced borosilicate glass to eliminate glass particles in the reduced sample.
Expand 2 Items
KIMBLE® DUALL® Tissue Grinders, All-Glass with Screw-Cap, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
The DUALL® is an extremely efficient grinder which combines both conical and cylindrical surfaces on a single pestle and tube. These separate areas perform the dual functions of initial grinding in the conical section and final homogenization in the cylindrical section.