Orders LinkContactUsLinkComponent
Dropper Bottles
Dropper Bottles
Dropper Bottles
Certificates
Dropper Bottles
3 Options Available Below
Product Details & Documents
These bottles are made of low-density polyethylene.
  • Bottles Dispense about 40 mcL per Drop
  • Good Chemical Resistance
  • Semi-Transparent Bottles
  • Polyethylene Construction
The semi-transparent bottles feature color-coded caps.
They have good chemical resistance and dispense about 40 mcL per drop.
Product Family Options
Product Information
  • Capacity
    Availability
    Price
  • 470211-800
    Dropper Bottles
    Dropper Bottles
    4 ml
    Log in for availability
    Log in for availability
    Case of 25
    Case of 25 - $119.00
    $119.00
    Case of 25
       $119.00
    Supplier Number: 2752-9125
    Specifications:
    More
    Specifications
    Cat. No.
    470211-800
  • 470211-796
    Dropper Bottles
    Dropper Bottles
    8 ml
    Log in for availability
    Log in for availability
    Case of 25
    Case of 25 - $129.00
    $129.00
    Case of 25
       $129.00
    Supplier Number: 2752-9025
    Specifications:
    More
    Specifications
    Cat. No.
    470211-796
  • 470211-798
    Dropper Bottles
    Dropper Bottles
    15 ml
    Log in for availability
    Log in for availability
    Case of 25
    Case of 25 - $132.00
    $132.00
    Case of 25
       $132.00
    Supplier Number: 2752-9050
    Specifications:
    More
    Specifications
    Cat. No.
    470211-798