ContactUsLinkComponent
Documenting a Crime Scene Kit
Documenting a Crime Scene Kit
  470024-256
 :  Avantor
undefined
Documenting a Crime Scene Kit
  470024-256
 :  Avantor
  • Description:
    Documenting a Crime Scene Kit
  • No. of Lab Groups:
    6
  • Cat. no.:
    470024-256
  • Supplier No.:
    WARD470024-256
Reinforce Math and Mapping Skills
  • Links to a real world application
  • Estimated class/lab time required: 45-60 minutes
Learn the systematic methods used by investigators as they conduct thorough searches and create illustrations. By performing a variety of crime scene search methods, several types of crime scene sketches can also be practiced. Strip, spiral, wheel, grid, and zone searches are addressed in addition to rough, final, cross projection, baseline, smooth, and street sketches. Includes materials for six setups.