DNA SuperModel Kit
DNA Supermodel Kit, 1
Catalog # 470029-868
Supplier: RYLER ENTERPRISES, INC.
Supplier #: DNA-1
Jump to:
Product Details & Documents
Re-Create Accurate Bonding Between Base Pairs
Easy to set up, this model is flexible, durable, and economical. The colorful components provide an accurate representation of bonding between base pairs by showing the double bonds between adenine and thymine and the triple bonds between guanine and cytosine. The kit comes with 183 pieces, which is enough to make a 24 nucleotide sequence. Includes an information and instruction booklet.
Caution: WARNING: CHOKING HAZARD - Small parts. Not for children under 3 yrs.
Specifications
- Description:DNA Supermodel Kit
- No. of lab groups:1