Orders LinkContactUsLinkComponent
LaMotte® Dissolved Oxygen Water Test Kit (Winkler Method)
LaMotte® Dissolved Oxygen Water Test Kit (Winkler Method)
Dissolved Oxygen Water Test Kit
Catalog # 470203-112
Supplier:   LaMotte
Supplier #:  5860-01
LaMotte® Dissolved Oxygen Water Test Kit (Winkler Method)
Catalog # 470203-112
Supplier:  LaMotte
LaMotte® Dissolved Oxygen Water Test Kit (Winkler Method)
LaMotte® Dissolved Oxygen Water Test Kit (Winkler Method)
Dissolved Oxygen Water Test Kit
Price
$126.35
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Precalibrated test tubes and labware give fast, accurate results on-site, in the classroom, or in the lab.
  • Range: 0 to 10 ppm
  • Materials for 50 tests
  • Aligned with GLOBE protocols
Reagents, labware, and step-by-step instructions are furnished in a compact, waterproof carrying case.
Documents
Specifications
  • Description:
    Dissolved Oxygen Water Test Kit
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)