Digestive System Slide Set
Digestive System Slide Set, 35, Animal
Catalog # 470183-602
Supplier: Avantor
Jump to:
Product Details & Documents
Various Organs, Glands, and Tissues Involved in the Digestive Process
- Identifying features are clearly distinguishable
- Rigorous quality control standards
Ordering information: Set contents include: Teeth (8 slides), Tongue (4 slides), Salivary Glands (2 slides), Soft Palate, Lip, Esophagus/Trachea (3 slides), Stomach (4 slides), Intestinal Area (8 slides), Pancreas, Gallbladder, and Liver (2 slides).
Specifications
- Type:Animal
- Description:Digestive System Slide Set
- No. of slides:35