Orders LinkContactUsLinkComponent
Diffraction Grating Glasses
Diffraction Grating Glasses
Diffraction Grating Glasses, 6
Catalog # 470015-970
Supplier #:  01101
Diffraction Grating Glasses
Catalog # 470015-970
Diffraction Grating Glasses
Diffraction Grating Glasses
Diffraction Grating Glasses, 6
Price
$20.20
Log in for availability
Pack of 1
Product Details & Documents
These glasses make the study of light and color fun and easy.
  • Double-Axis Glasses
  • Lenses Separate Light from Other Sources
  • Economical
13,500 lines per inch double-axis holographic grating lenses separate light from any source into its spectral components for study and analysis.
Glasses are priced inexpensively enough to outfit your entire class.
Packaging: One package contains six glasses.
Specifications
  • Description:
    Diffraction Grating Glasses
  • No. of parts:
    6