If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Description:WARD’S Determining Vitamin C Concentration Lab Activity
- Cat. no.:470004-922
- Supplier No.:WARD470004-922
Ideal to determine the amount of Vitamin C in fruits and vegetables.
Scientists are discovering that vitamin C plays a major role in human physiology. It is a key component in the formation of collagen, bones, and teeth, and it aids in fighting infection. In this interesting activity, students first titrate a known, standard solution of vitamin C. They then prepare their own samples using common fruits and vegetables (fresh, canned, or frozen) that you supply. By comparing their results to the standard solution, students can quantify the amount of vitamin C present in their own samples. It includes enough materials for eight setups, a teacher’s guide, and student study guide copymaster.