ContactUsLinkComponent
Density Tube
Density Tube
Catalog # 470152-828
Supplier:  Avantor
undefined
Density Tube
Catalog # 470152-828
Supplier:  Avantor
Specifications
  • Description:
    Density Tube
  • Cat. no.:
    470152-828
  • Supplier No.:
    WARD470152-828
Specifications
About this item
Teacher Developed!
  • Grades 7–12
  • Size: 24.8cm H
Physics teacher Victor Noto has developed a great way to demonstrate heat transfer by convection. His density tube uses water at different temperatures to produce bulk liquid flow. By timing how long it takes for a small volume of liquid to move up to the top of the tube, students can determine if the rate of heat transfer depends on the density differences. Includes instructions.