Orders LinkContactUsLinkComponent
Density Cylinder Set
Density Cylinder Set
Cylinder, Density Set, Set/4
Catalog # 470138-850
Supplier:   Wards
Supplier #:  PH0112F
Density Cylinder Set
Catalog # 470138-850
Supplier:  Wards
Density Cylinder Set
Density Cylinder Set
Cylinder, Density Set, Set/4
Price
$52.35
Log in for availability
Kit of 1
Product Details & Documents
This Equal Mass Density Set is for specific gravity and density determinations.
  • For Specific Gravity and Density Determinations
  • Four Cylinders of Varying Volumes Included
  • Width of All Cylinders: 16 mm dia.
  • Consist of: Cu, Fe, Al, Zn
It's used to study the specific relationship between mass and density.
The set includes four cylinders with the same mass and diameter (16 mm) but varying volumes.
Specifications
  • Description:
    Cylinder, Density Set, Set/4