Demonstration Balance
Catalog # 470104-170
Supplier: Avantor
Specifications
- Description:Balance Demonstration (without meter stick)
- Cat. no.:470104-170
- Supplier No.:WARD470104-170
Specifications
About this item
Inexpensive but Important Apparatus
- One knife-edge clamp
- Heavy cast metal support
For demonstrating the principle of moments. A meter stick (not supplied) is easily attached using a set screw fitted on the clamp. Complete with instructions and suggestions for use.