If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Description:Decomposition Lab Activity
- No. of Lab Groups:50
- Cat. no.:470039-840
- Supplier No.:35-A
Specifications
About this item
Guided Inquiry Study of Soil
- Students will create compost
- Demonstrate biodegradability
- Connects with other environmental concepts
Explore the concepts of biodegradability and decomposition and how they apply to the study of life cycles. This series of inquiry-based experiments investigate the effect of soil type on decomposition, the different types of substances which decompose, and demonstrate how to determine the rate of decomposition. Includes materials for 50 students.