Order Entry
Canada
ContactUsLinkComponent
 
Customary SAFE-T Weights™ Set
  470046-274
undefined
Customary SAFE-T Weights™ Set
  470046-274
 :  32060

 

  • Description:
    Set, Weight, 13 Piece Customary, (Total = 3 lbs), Safe-T Weights
  • Cat. no.:
    470046-274
  • Supplier No.:
    32060

 

 

The U.S. Customary (or English) SAFE-T Weights™ have the same features as the Metric SAFE-T Weights™.

  • Understand two measuring systems
  • Same features as metric version

By comparing our 1 pound weight to the metric 1,000 gram (or kilogram), students can quickly develop the feel or understanding of these two different systems of measure. Contains eight 1 oz., 2 each of 4 oz. and 8 oz., and one 1 lb. weights, for a total weight of 3 lb.

 : WARNING: CHOKING HAZARD- Small Parts. Not for Children Under 3 Years.