Crystal Lattice Models
Crystal Lattice Models, Complete Set of 9 Models
Catalog # 470123-054
Supplier: Klinger
Supplier #: KS7996SPL
Jump to:
Product Details & Documents
Choose one, or all of our specific structures models to introduce concepts of unit cells, coordination numbers and crystal systems; or as valuable aids when discussing solid state, X-ray diffraction, cohesive forces and other topics. (The characteristic unit cell can be indicated by colored thread or colored connecting rods).
- The models comprise 1", color-coded, wooden spheres attached by sturdy metal rods
- Dimensions along each edge vary from 4 to 9", depending upon the structure represented
- Each model is furnished completely assembled
A comprehensive, illustrated teacher's guide explains the fundamental crystal systems and discusses the use of each model.
Specifications
- Description:Crystal Lattice Models, Complete Set of 9 Models
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



