Orders LinkContactUsLinkComponent
Crop Identification Kit
Crop Identification Kit
Crop Identification Kit
Catalog # 470004-846
Supplier:   Young Naturalist
Supplier #:  205
Crop Identification Kit
Catalog # 470004-846
Supplier:  Young Naturalist
Crop Identification Kit
Crop Identification Kit
Crop Identification Kit
Price
$103.75
Log in for availability
Kit of 1
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Dehydrated Preserved
This hands-on study looks at ten important crops and their by-products. It includes ten real crops that are each carded, labeled, and bagged; and ten crop by-products; as well as a teacher’s guide. Set of 20.
Specifications
  • Description:
    Crop Identification Kit
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)