Order Entry
Canada
ContactUsLinkComponent
 
Counter Brush
  470203-396
 :  Mill-Rose
undefined
Counter Brush
  470203-396
 :  Mill-Rose
 :  9090M

 

  • Bristle Material:
    Horse hair
  • Description:
    Counter Brush
  • Total length:
    14 1/2"
  • Environmentally Preferable:
  • Cat. no.:
    470203-396
  • Supplier No.:
    70035
  • Brush Head Length:
    8"
  • Trim Length:
    2"

 

 

Constructed of Horse Hair and Tampico Bristles.