Bottles, Square, Polycarbonate, Corning
Supplier: Corning
Jump to:
About this item
PC, square.
- More break-resistant than other glass or plastic bottles
- Screened white enamel graduations are easier to see than moulded graduations
- Large white marking spot for easier identification
- Bottles can be autoclaved once at 121 °C and 15 psi (repeated autoclaving weakens PC and is not recommended)
- Can be stored at –80 °C
These storage bottles are easier to handle, require less space on the shelf or in the autoclave, and are ideal for mixing, sampling and storage.
3 Options Available Below
Product Details & Documents
PC, square.
- More break-resistant than other glass or plastic bottles
- Screened white enamel graduations are easier to see than moulded graduations
- Large white marking spot for easier identification
- Bottles can be autoclaved once at 121 °C and 15 psi (repeated autoclaving weakens PC and is not recommended)
- Can be stored at –80 °C
These storage bottles are easier to handle, require less space on the shelf or in the autoclave, and are ideal for mixing, sampling and storage.
Documents
Certificates
Search by Lot NumberProduct Family Options
Product Information
- AvailabilityPrice
- 470211-300Corning Square Polycarbonate Bottles, 250 mLCorning Square Polycarbonate Bottles, 250 mLEach$13.60Case of 24$326.40Supplier Number: 431431
- 470211-302Corning Square Polycarbonate Bottles, 500 mLCorning Square Polycarbonate Bottles, 500 mLEach$17.00Case of 24$408.00Supplier Number: 431432
- 470211-304Corning Square Polycarbonate Bottles, 1000 mLCorning Square Polycarbonate Bottles, 1000 mLEach$22.70Case of 24$544.80Supplier Number: 431433
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.