Orders LinkContactUsLinkComponent
Coolant Trap
Coolant Trap
Coolant Trap
Catalog # 470022-252
Supplier:   GM DIECRON, INC.
Supplier #:  28-1921
Coolant Trap
Catalog # 470022-252
Supplier:  GM DIECRON, INC.
Coolant Trap
Coolant Trap
Price
$2,595.45
Log in for availability
Each
Product Details & Documents
Extends pump life.
  • Keeps coolant and other debris from entering the vacuum pump
  • Suitable for all thin section saws/grinders sold by WARD’S
Specifications
  • Description:
    Coolant Trap