Concepts of Light Classroom Kit
Catalog # 470148-940
Supplier: Shiv Dial Sud & Sons
Specifications
- Description:Concepts of Light, Classroom Version
- Cat. no.:470148-940
- Supplier No.:PH30912A
Specifications
About this item
Help Your Students Perceive How Light Behaves
Using mirrors, lenses, and a ray box, you can help your students better understand how light can be bent or reflected in different ways based on the shape of the lens or mirror. You can also explore how light and color are related. Kit includes an acrylic prism set with a triangular, half-round, double convex, and rectangular prism and two double concave prisms all packed in a wooden storage box; a ray box; Newton’s color disk; a plane and concave mirror; and instructions with correlations to the National Science Education Standards, student instruction sheets, and suggestions for science fairs.
Ordering information: A 12V DC 3A power supply is required to run this ray box. Power supply not included.