If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Description:LaMotte® GREEN Comprehensive Water Monitoring Kit
- No. of Lab Groups:100
- Cat. no.:470004-240
- Supplier No.:5848
Most factors can be tested 100 times each with the Comprehensive Water Monitoring Kit.
- No hazardous compounds needed for testing
- Monitoring kit manual is in english and spanish
- Single-factor test kits
- Enough TesTabs to test nine factors 100 times each
- Aligned to NGSS 4-ESS2-1, 4-ESS2-2, MS-LS2-1, MS-LS2-5, MS-ESS3-3, HS-LS2-7, HS-ESS2-5, HS-ESS3-4
Using TesTabs, which are safe, nonhazardous reagent tablets, students can easily test for a variety of factors without risk. Developed by the Global Rivers Environmental Education Network, each monitoring kit comes with a bilingual (English and Spanish) manual.
The kit include enough TesTabs to test nine factors. Test factors include benthic macroinvertebrates, biochemical oxygen demand, coliform bacteria, nitrate, dissolved oxygen, pH, phosphate, turbidity, and temperature change. The kit have enough material for 100 tests each (44 coliform tests; unlimited benthic invertebrate, turbidity, and temperature change tests).