Colon, Mammal, Thin Section Slide
Catalog # 470183-688
Supplier: Avantor
Specifications
- Description:Colon, Mammal, Thin Section
- Cat. no.:470183-688
- Stain:Hematoxylin and Eosin
- Supplier No.:WARD470183-688
- Orientation:Thin section
Specifications
About this item
Extremely Thin Plastic Embedded Tissue For Greater Viewing Clarity
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. Through the large intestine, showing absence of villi and changes in muscular layers. Goblet cells are numerous.
Through the large intestine, showing absence of villi and changes in muscular layers. Goblet cells are numerous.