Collision Ball Apparatus
470005-398
: Wards
- Description:Collision Ball Apparatus
- Cat. no.:470005-398
- Supplier No.:COL-12
An easy, inexpensive way to effectively demonstrate conservation of energy and momentum
- Durable
- High Quality
This apparatus includes five steel balls made to collide with each other on a metal track. The metal track is 58 cm long and is supported at both ends.