Coliform Test Kit
Less confusing than other test kits that require students to distinguish different bacterial colonies on a Petri dish, this test features a distinct color change that will tell with certainty if fecal coliforms are present.
- Students Determine the Presence or Absence of Harmful Bacteria
- Simple to Use and Interpret
- Instructions Included
- Material for 25 Tests
- Qualitative Coliform Test Yields Results within 24 Hours
The marked color change determined by E. coli presence occurs overnight in a lactose broth.
It includes materials for 25 tests and instructions.
- Description:Coliform Test Kit
- No. of lab groups:25
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)