ContactUsLinkComponent
Cobalt Chloride Test Paper
Cobalt Chloride Test Paper
Catalog # 470123-112
undefined
Cobalt Chloride Test Paper
Catalog # 470123-112
Supplier Number:  150-12V-100
Specifications
  • Description:
    Cobalt Chloride Test Paper
  • Cat. no.:
    470123-112
  • Supplier No.:
    150-12V-100
  • Quantity Per Box/Pack/Carton:
    12 vials
  • No. of Strips:
    100
Specifications
About this item
Cobalt Chloride cloth detects the presence of moisture to determine if you have wetness or dampness problems.
The Cobalt chloride cloth is responsive to 50% relative humidity and above, and any direct measure of an aqueous solution.
Packaging: 12 Vials of 100 Strips.