Clostridium rubrum Slide
470180-894
: Avantor
- Description:Clostridium rubrum slide
- Cat. no.:470180-894
- Stain:Gram (+)
- Supplier No.:WARD470180-894
- Orientation:Smear
Demonstrates Well-defined Endospore
- Identifying features clearly distinguishable
- Rigorous quality control standards
An aerobic, spore forming, rod-shaped bacterium. Spores are oval in shape, and subterminal in location within the cell. This sometimes makes the cells appear to be ‘fatter’ at one end than the other.