Clay Triangles
Clay Triangles, 2"
470176-154
: CPT020
Clay pipe triangle for holding items over a flame.
- Heat resistant clay pipes
Galvanized, 16-gauge iron wire with plain clay-pipe stems.
- Description:Clay Triangles, 2"