ContactUsLinkComponent
Chemical Formulas Lab Activity
  470200-084
 :  Kemtec
undefined
Chemical Formulas Lab Activity
  470200-084
 :  Kemtec
 :  13-122
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    Chemical Formulas Lab Activity
  • Environmentally Preferable:
  • Cat. no.:
    470200-084
  • Supplier No.:
    13-122
Experimentally Determine the Correct Chemical Formulas for Various Compounds
By combining chemicals in appropriate ratios, students can determine the correct chemical formula of a new compound by observing the combination that produces the greatest amount of precipitate. Using the ratio of the combination, they can determine the chemical formula for the new substance formed. You will receive enough materials for four setups and reproducible lab and assessment manuals.