If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Description:Characteristics of Matter Lab Activity
- No. of Lab Groups:6
- Cat. no.:470100-890
- Includes:Instructors Manual and Student Data Sheets
- Supplier No.:1-114
Determine the Many Properties of Matter
- Meets national standards for grades 6-9
- Combine scientific inquiry with structured assessments
Ten experiments in which students differentiate between physical and chemical properties and explore the characteristics of matter; color, odor, solubility, shape, size, texture, density, alkalinity, acidity, states of matter, boiling point, and melting point. Includes graphing of melting and boiling points. Students observe chemical properties of five solutions by observing their reactions with other solutions. From these reactions, students then determine the identity of two unknown solutions. Includes reproducible student data sheets, assessment questions, and answer keys. Required, but not included: goggles, balance, ice, test tubes, beaker or clear cup, 10 mL graduated cylinder, and test tube racks.
Includes:
Instructor Manual
Spot Plates
Wood Stirrers
Thermometers
Plastic Bottles with Droppers & Caps
Blue Litmus
Red Litmus
Pipettes
Ammonia Solution
Baking Soda Solution
Calcium Chloride Solution
Vinegar
Washing Soda Solution
Calcium Chloride Solution
Calcium Carbonate
Sodium Chloride
Sand
Sucrose