Cell Nucleus Microslide
About this item
Explore diverse cell types
- Arrows help students locate important features
- Mounted in a clear plastic holder that protects it on both sides
- Set of 15 includes detailed lesson plan on the topic under study
Microslides consist of a strip of film with eight photomicrographs, each reproduced exactly as you would see them through a professional microscope under ideal conditions. You can supply all your students with their own Microslides. A detailed text folder is included. Viewers are available separately.
Contents:
Animal cell nucleus (900X)
Plant cell nucleus (1,000X)
Nuclear reproduction (100X)
Human blood cells (900X)
Voluntary muscle cells (900X)
Nuclear membrane (em, 100,000X)
Nuclear membrane surface (em, 100,000X)
Endoplasmic reticulum (em, 125,000X)
2 Options Available Below
Product Details & Documents
Explore diverse cell types
- Arrows help students locate important features
- Mounted in a clear plastic holder that protects it on both sides
- Set of 15 includes detailed lesson plan on the topic under study
Microslides consist of a strip of film with eight photomicrographs, each reproduced exactly as you would see them through a professional microscope under ideal conditions. You can supply all your students with their own Microslides. A detailed text folder is included. Viewers are available separately.
Contents:
Animal cell nucleus (900X)
Plant cell nucleus (1,000X)
Nuclear reproduction (100X)
Human blood cells (900X)
Voluntary muscle cells (900X)
Nuclear membrane (em, 100,000X)
Nuclear membrane surface (em, 100,000X)
Endoplasmic reticulum (em, 125,000X)
Product Family Options
Product Information
- AvailabilityPrice
- 470009-116Cell nucleus microslideSupplier Number: T-248
- 470022-044Cell nucleus microslide, set of 15Supplier Number: T-248-15B