Cell Division Slide Set
Cell Division Slide Set, 2, Cellular Development
Catalog # 470183-556
Supplier: Avantor
Jump to:
Product Details & Documents
Unique Comparison of Plant and Animal Mitosis
- Identifying features clearly distinguishable
- Rigorous quality control standards
Ordering information: Set includes Plant (onion) mitosis (ls); and Animal (whitefish blastodisc) mitosis (sect).
Specifications
- Type:Cellular Development
- Description:Cell Division Slide Set
- No. of slides:2