Calibration Thermometer
Calibration Thermometer, −30…+50 °C
Catalog # 470001-916
Supplier: Thermco
Supplier #: BCP2019PS
Jump to:
Product Details & Documents
Calibrated for Greatest Accuracy
The durable, long lasting plastic armor covering this thermometer includes an attachment loop. It is also ideal for use as part of the GLOBE program. Total immersion. Red alcohol. 6"L
Specifications
- Description:Calibration Thermometer
- Range:−30…+50 °C