C Clamps
About this item
Securely hold two flat object together.
- Cast iron frame
- Screw thread mechanism
These cast iron C clamps help to securely holes surfaces together. The maximum opening of the jaw needs to be taken into account when selecting the correct size. The horizontal torque bar allows you to apply a lot of force to tighten or release.
2 Options Available Below
Product Details & Documents
Securely hold two flat object together.
- Cast iron frame
- Screw thread mechanism
These cast iron C clamps help to securely holes surfaces together. The maximum opening of the jaw needs to be taken into account when selecting the correct size. The horizontal torque bar allows you to apply a lot of force to tighten or release.
Product Family Options
Product Information
- AvailabilityPrice
- 470005-3688 cm (3") Jaw OpeningSupplier Number: CLMPC3Specifications:
More
Specifications
Cat. No.470005-368 - 470104-09410 cm (4") Jaw OpeningSupplier Number: CLMPC4Specifications:
More
Specifications
Cat. No.470104-094