Polypropylene Buret Clamp
Catalog # 470144-830
Supplier: United Scientific Supplies
Specifications
- Description:Polypropylene Buret Clamp, Single
- Cat. no.:470144-830
- Supplier No.:45101
Specifications
About this item
Non-corroding, polypropylene plastic clamps hold burettes firmly in place.
- Corrosion-proof
- Spring loaded to hold burettes
These polypropylene burette clamps are completely corrosion-proof. The spring-loaded jack in the middle provides a vice like grip on the burette and prevents it from slipping. Two tapered wedges are included to ensure firm grip on 1/2" or 3/8" diameter rods.