Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Buffer Envelopes

Buffer Envelopes
 
Buffer Envelopes
Buffer Envelopes

 

 
   
 

Shelf Life (months): 36
Storage: Green

 : Please Note: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Product Family Options

 

  • Description
    Grade
    Chemical/physical specifications
    Size
     
     
  • Buffer Envelope
    Laboratory
    pH 3.00
    Pack of 5
       
       
    Pack of 5
    Pack of 5 - $34.45
    $34.45
    Pack of 5
       $34.45

     : 280-PH3
  • Buffer Envelope
    Laboratory
    pH 6.00
    Pack of 5
       
       
    Pack of 5
    Pack of 5 - $34.45
    $34.45
    Pack of 5
       $34.45

     : 280-PH6
  • Buffer Envelope
    Laboratory
    pH 8.00
    Pack of 5
       
       
    Pack of 5
    Pack of 5 - $34.45
    $34.45
    Pack of 5
       $34.45

     : 280-PH8
  • Buffer Envelope
    Laboratory
    pH 11.00
    Pack of 5
       
       
    Pack of 5
    Pack of 5 - $34.45
    $34.45
    Pack of 5
       $34.45

     : 280-PH11
  • Buffer Envelope
    Laboratory
    pH 12.00
    Pack of 5
     
     : -

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.