Brevibacillus brevis
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
All microbial cultures undergo an extensive quality control program prior to releasing for sale, so they are guaranteed for purity as well as for species.
- Biosafety Level: 1
- Gram Stain: Positive
- Cell Morphology: Rods
- Respiration: Aerobic
Ward’s Science is not affiliated with the American Type Culture Collection and does not provide ATCC numbers for our organisms.
Ordering information: Microbial and fungal cultures are restricted and can only be ordered by educational professionals.
These products contain living or perishable materials and will ship via overnight or second day delivery. Please specify a delivery date during checkout
Caution: While this organism is non-pathogenic, Ward’s recommends that you utilize proper aseptic technique when handling all microorganisms.
Documents
Bacteria_Fungi Working_with_Bacteria_and_Fungi
Product Literature
Specifications
- Description:B. brevis
- Storage conditions:Refrigerator
- Packaging:Tube
- Media:Azotobacterial agar
- Incubation conditions:30 °C
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



