ContactUsLinkComponent
Ward's® Prepared Brain-Heart Infusion Agar
Ward's® Prepared Brain-Heart Infusion Agar
Catalog #: 470180-706
CAS Number:  
PLATES BRAIN-HEART INFUSION AGAR PK/10
Ward's® Prepared Brain-Heart Infusion Agar
Catalog #: 470180-706
CAS Number:  
Specifications
  • Packaging:
    Plates
  • Description:
    Prepared Brain-Heart Infusion Agar
  • Cat. no.:
    470180-706
  • Supplier No.:
    WARD470180-706
Specifications
About this item
Brain-Heart Infusion (BHI) Agar is a general purpose media used for the cultivation of fastidous microrganisms.
Excellent media for cultivaion of such as streptococci, pneumococci and meningococci.
Ordering information: These products contain living or perishable materials
Delivery: Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.
Customers Who Bought This Also Bought