Orders LinkContactUsLinkComponent
Box Magnifiers
Box Magnifiers
Box Magnifiers
Certificates
Box Magnifiers
2 Options Available Below
Product Details & Documents
Contains and Magnifies Small Specimens
These clear plastic boxes feature a magnifying press-fit lid; simply place the specimen to be studied in the box, cover, and observe. They can also be used when permanently embedding small specimens in resin. Size: 4X magnifier 1 L×1 W×7/8H"; 3X magnifier 1 1/2 L×1 1/2 W×1 1/2 H".
Product Family Options
Product Information
  • Magnification
    Availability
    Price
  • 470016-876
    Box Magnifier
    Box Magnifier
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $4.90
    $4.90
    Log in for availability
    Box of 250
    Box of 250 - $1,169.20
    $1,169.20
    Each
       $4.90
    Box of 250
       $1,169.20
    Supplier Number: 6455000
  • 470016-878
    Box Magnifier
    Box Magnifier
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $7.90
    $7.90
    Log in for availability
    Box of 250
    Box of 250 - $1,898.55
    $1,898.55
    Each
       $7.90
    Box of 250
       $1,898.55
    Supplier Number: 6455100