Ward's® Bocce Ball Pendulum
Bocce Ball Pendulum
Catalog # 470039-722
Supplier: Wards
Supplier #: 180209
Jump to:
Product Details & Documents
Large scale demonstration of harmonic motion.
- Large scale demonstration
- Easily attaches to any ceiling
Help students see the answers to conservation of energy, vectors, and tension questions are right in front of their faces. This classical experiment has been adapted to excite, engage, and intrigue students. A 5 lb. bocce ball is hung from a 25 ft. long rope with a breaking point of 500 lbs. Students hold this ball to their nose and release it, with the goal to stay still as it returns just short of its initial position. For extra stability, the rope can be pulled through the ball and be hung by two ropes instead of one.
Specifications
- Description:Bocce Ball Pendulum