SK Borosilicate Graduated Cylinders
Catalog #: 470104-152
Supplier: Avantor
Specifications
- Description:Tall form cylinder, SK student, borosilicate, double scale, 25×0.5 ml
Specifications
About this item
Sturdy graduated cylinders for classroom use.
- Cylinders comply with ASTM E1272, Class B standards
- Stable hexagon base
These fine quality, student grade cylinders have round bases with heavy fire-polished rims and pour-outs.
The graduations are white lined with the principal lines slightly longer. Calibration is adjusted to 20°C. Available in both single and double scale in the sizes below.
Customers Who Bought This Also Bought