PYREX® Round Bottom Boiling Flasks
470202-168
: Corning
Product available on GSA Advantage®
- Description:Flask, boiling, Pyrex®, 1000 ml, w/ round bottom, Corning
- Environmentally Preferable:
- Cat. no.:470202-168
- Supplier No.:4280-1L
Recommended for critical boiling applications.
These round bottom flasks are made from quality borosilicate and have a heavy neck rim to minimize breakage.