ContactUsLinkComponent
Blood Typing Using Saliva Lab Activity
Blood Typing Using Saliva Lab Activity
Catalog # 470007-746
Supplier:  Kemtec
undefined
Blood Typing Using Saliva Lab Activity
Catalog # 470007-746
Supplier:  Kemtec
Supplier Number:  3-214
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Blood Typing Using Saliva Lab Activity
  • Environmentally Preferable:
  • No. of Lab Groups:
    24
  • Cat. no.:
    470007-746
  • Supplier No.:
    3-214
Specifications
About this item
This kit lets students determine their blood types without drawing a single drop of blood.
  • Material for 24 Students
  • Saliva Determines Antigens and Blood Type
  • No Blood Drawing Needed
  • Centrifuge Required
Students use their saliva to determine if they are among the 80% of the population who secrete antigens. Those who are then determine their ABO blood type.
The kit includes material for 24 students. A centrifuge is required but not included.
Ordering information: This item contains a coupon for perishable materials that must be redeemed from the manufacturer before use. Please allow sufficient time to account for redemption. These materials are shipped from the supplier via 2nd day delivery on Mondays, Tuesdays, or Wednesdays only and must be received promptly.