ContactUsLinkComponent
Ward's® Bioremediation of Industrial Pollution Lab Activity
Ward's® Bioremediation of Industrial Pollution Lab Activity
Catalogue # 470030-304
Fournisseur:  Avantor
undefined
Ward's® Bioremediation of Industrial Pollution Lab Activity
Catalogue # 470030-304
Fournisseur:  Avantor
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Ward's® Bioremediation of Industrial Pollution Lab Activity
  • No. of Lab Groups:
    6
  • Cat. no.:
    470030-304
Spécifications
À propos de cet article
Re-create “Pollution” and Observe Bacteria’s Degradation Abilities
  • Linked to a real world scenario
  • Includes a teacher's guide
  • Estimated class/lab time required: 90 minutes over one lab session and three follow-up sessions
Students will be able to study bioremediation, a process that uses naturally occurring organisms to degrade hazardous waste into nontoxic materials, and assess its potential as a viable method of cleaning waste sites with our unique lab activity. In an investigation that spans one 45-minute lab session and three brief, 15-minute follow-up sessions, students grow a naturally occurring bacterial strain and observe its degrading properties on completely safe, simulated “chemical waste”.
Ordering information: Includes enough materials for six setups, a teacher’s guide, and student copymaster. An incubator, required, is available separately. Coupon included for perishable materials. Redeem by phone or e-mail.