Order Entry
Canada
ContactUsLinkComponent
Bimetallic Dial Thermometers with Clip-on Pocket Case
Bimetallic Dial Thermometers with Clip-on Pocket Case
Catalog # 470204-674
Supplier:  Bel-Art Products
undefined
Bimetallic Dial Thermometers with Clip-on Pocket Case
Catalog # 470204-674
Supplier:  Bel-Art Products
Supplier Number:  B61310-3400

Specifications

  • Description:
    Bimetallic Dial Thermometers with Clip-on Pocket Case
  • Cat. no.:
    470204-674
  • Dial Diameter:
    1-3/8"
  • Divisions:
    5 °F/2 °C
  • Range:
    0...220 °F / −10...100 °C
  • Supplier No.:
    21125

Specifications

About this item

These dial thermometers feature superior shock protection, easy readability and external recalibration.

The stem is clad in stainless steel and the dial is covered with an unbreakable polycarbonate crystal.

Stem length: 5".

Delivery: Nylon pocket case included.